Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MS(PEG)12 Methyl-PEG-NHS-Ester Reagent

Catalog No. p-7184460
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22685 100 mg
PI22686 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI22685 Supplier Thermo Scientific™ Supplier No. 22685
Only null left
Add to Cart
Edge
Add to Cart

Pegylate amines (-NH2 groups) on proteins (lysine residues) or assay surfaces using MS(PEG)n, amine-reactive (NHS ester), methyl-capped PEG compounds (4 to 24 EG units).

Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.

MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.

Highlights:

  • NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
  • Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Easy-to-follow instructions increase the likelihood of a successful outcome
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Content And Storage Store desiccated at -20°C.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer 12 ethylene glycol units
Form Solid
Quantity 100 mg
Solubility Water
Product Type NHS-Ester Reagent

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.