Learn More
Laxco™ MS Series Fixed Magnification Stereo Microscope System
SDP
Utilizes fixed dual magnification optical system. Laxco™ MS Series Fixed Magnification Stereo Microscope System are research-grade instrument with wide choice of zoom magnification ranges and most commonly found in general laboratory.
Specifications
Warranty 2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Product Type Stereo Microscope
Lighting Type Transmitted Fluorescent and Halogen Reflected
View More Specs
Products 4
Catalog Number Mfr. No. Magnification Power Eyepiece Price Quantity & Availability  
Catalog Number Mfr. No. Magnification Power Eyepiece Price Quantity & Availability  
MS20420
SDP
View Documents
Laxco™
MS20420
40X to 80X 20X N/A N/A N/A
MS204
SDP
View Documents
Laxco™
MS204
20X to 40X 10X N/A N/A N/A
MS20415
SDP
View Documents
Laxco™
MS20415
15X to 45X 15X N/A N/A N/A
MS103
SDP
View Documents
Laxco™
MS103
10X to 30X 10X N/A N/A N/A
Description
Description
Optics
  • Choose between multiple combinations of magnifications
  • Dual diopter adjustments accommodates different eye focal length
  • Parfocality maintained between magnification
  • Clearly marked neural knobs for magnification
Stand
  • Rugged, all metal construction
  • Large, contoured stage
  • Left and right hand operation
  • Slip clutch prevents damage to the focusing mechanism
  • Locked-in stage clip
Light Source
  • 20w halogen lower illumination
  • 15w halogen, with reflector for upper illumination
  • Individually control light sources
Eyepiece
  • 10X super-widefield eyepieces with interpupillary and diopter adjustments to correct for user-specific eyesight
Certifications
Quality Proformance Certification
Specifications
Specifications
2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Stereo Microscope
Transmitted Fluorescent and Halogen Reflected
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.