Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mS2i6A Mouse anti-Mammalia, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABS129525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1295-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1295-25 Supplier MilliporeSigma Supplier No. MABS129525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MS2i6A, clone CL-3, Cat. No. MABS1295, is a mouse mouse monoclonal antibody that detects mS2i6A and has been tested for use in Dot Blot, ELISA, Immunocytochemistry, and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mS2i6A
Applications Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen A synthesized mS2i6A oligonucleotide conjugated to BSA.
Molecular Weight of Antigen N/A
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 3 (CL-3) recognizes wild-type total RNA but not knockout total RNA.
Target Species Mammalia
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.