Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A12 Antibody (OTI5F5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272791H
Description
MS4A12 Monoclonal specifically detects MS4A12 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MS4A12 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MS4A12 | |
Full length recombinant protein of human MS4A12 (NP_060186) produced in HEK293T cells. | |
0.1 mL | |
54860.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5F5 | |
Western Blot, Immunohistochemistry | |
FLJ20217, membrane-spanning 4-domains subfamily A member 12, membrane-spanning 4-domains, subfamily A, member 12, Ms4a10 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction