Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A12 Monoclonal Antibody (4F4)
GREENER_CHOICE

Catalog No. PIMA524481
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524481 Supplier Invitrogen™ Supplier No. MA524481
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MMSSKPTSHA EVNETIPNPY PPSSFMAPGF QQPLGSINLE NQAQGAQRAQ PYGITSPGIF ASSQPGQGNI QMINPSVGTA VMNFKEEAKA LGVIQIMVGL MHIGFGIVLC LISFSFREVL GFASTAVIGG YPFWGGLSFI ISGSLSVSAS KELSRCLVKG SLGMNIVSSI LAFIGVILLL VDMCINGVAG QDYWAVLSGK GISATLMIFS LLEFFVACAT AHFANQANTT TNMSVLVIPN MYESNPVTPA SSSAPPRCNN YSANAPK.

The protein encoded by this gene is a cell surface protein found primarily in the apical membrane of colonocytes. Silencing of this gene in colon cancer cells inhibits the proliferation, cell motility, and chemotactic invasion of cells. This gene is part of a cluster of similar genes found on chromosome 11. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A12
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MS4A12
Gene Accession No. Q9NXJ0
Gene Alias membrane spanning 4-domains A12; membrane-spanning 4-domains subfamily A member 12; membrane-spanning 4-domains, subfamily A, member 12; Ms4a10; MS4A12
Gene Symbols MS4A12
Host Species Mouse
Immunogen MS4A12 (AAH29793, 1 a.a. ~ 267 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54860
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.