Learn More
MS4A12 Mouse anti-Human, Clone: OTI5F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808071

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a cell surface protein found primarily in the apical membrane of colonocytes. Silencing of this gene in colon cancer cells inhibits the proliferation, cell motility, and chemotactic invasion of cells. This gene is part of a cluster of similar genes found on chromosome 11. Two transcript variants encoding different isoforms have been found for this gene.Specifications
MS4A12 | |
Monoclonal | |
Unconjugated | |
MS4A12 | |
Ms4a10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
54860 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NXJ0 | |
MS4A12 | |
Full length human recombit protein of human MS4A12 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.