Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A15 Monoclonal Antibody (1D4)
GREENER_CHOICE

Catalog No. PIMA523183
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523183 Supplier Invitrogen™ Supplier No. MA523183
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MVRRGHVGIF FIEGGVPFWG GACFIISGSL SVAAEKNHTS CLVRSSLGTN ILSVMAAFAG TAILLMDFGV TNRDVDRGYL AVLTIFTVLE FFTAVIAMHF GCQAIHAQAS APVIFLPNAF SADFNIPSPA ASAPPAYDNV AYAQGVV.

May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A15
Applications ELISA
Classification Monoclonal
Clone 1D4
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MS4A15
Gene Accession No. Q8N5U1
Gene Alias membrane spanning 4-domains A15; membrane-spanning 4-domains subfamily A member 15; membrane-spanning 4-domains, subfamily A, member 15; MS4A15
Gene Symbols MS4A15
Host Species Mouse
Immunogen MS4A15 (AAH31610.1, 1 a.a. ~ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219995
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.