Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983082
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3082 Supplier Bioss Supplier No. BS13691R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in signal transduction as a component of a multimeric receptor complex.

Specifications

Antigen MS4A15
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MS4A15
Gene Accession No. Q8N5U1
Gene Alias membrane spanning 4-domains A15; Membrane-spanning 4-domains subfamily A member 15; membrane-spanning 4-domains, subfamily A, member 15; MS4A15
Gene Symbols MS4A15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MS4A15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219995
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.