Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560931 Supplier Invitrogen™ Supplier No. PA560931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VWAAKDPSPC VVNSSISFNI ISALF Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 60%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MS4A18
Gene Accession No. Q3C1V0
Gene Alias membrane spanning 4-domains A18; membrane-spanning 4-domains subfamily A member 18; membrane-spanning 4-domains, subfamily A, member 18; MS4A18
Gene Symbols MS4A18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MS4A18. Recombinant protein control fragment (Product #RP-100337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728588
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.