Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563806 Supplier Invitrogen™ Supplier No. PA563806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EELKGNKVPE DRVYEELNIY SATYSELEDP GEMSPPID Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 53%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MS4A2
Gene Accession No. Q01362
Gene Alias APY; ATOPY; beta polypeptide); fc epsilon receptor I beta-chain; Fc fragment of IgE, high affinity I, receptor for; beta polypeptide; Fc receptor, IgE, high affinity I, beta polypeptide; Fce1b; Fcer1b; fcERI; FCIGA; FcRB; Fcrbeta; high affinity IgE receptor beta subunit; High affinity immunoglobulin epsilon receptor beta-subunit (FcERI) (IgE Fc receptor, beta-subunit) (Fc epsilon receptor I beta-chain); high affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; immunoglobulin E receptor, high affinity, beta polypeptide; membrane spanning 4-domains A2; Membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains subfamily A member 2; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; membrane-spanning 4-domains, subfamily A, member 2 (Fc fragment of IgE, high affinity I, receptor for; membrane-spanning 4-domains, subfamily A, member 2 (Fc fragment of IgE, high affinity I, receptor for; beta polypeptide); MS4A1; Ms4a2
Gene Symbols MS4A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MS4A2. Recombinant protein control fragment (Product #RP-101361).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2206
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.