Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107183
Description
MS4A3 Polyclonal Antibody for Western Blot
This gene encodes a member of the membrane-spanning 4A gene family. Members of this protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This family member likely plays a role in signal transduction and may function as a subunit associated with receptor complexes. The gene encoding this protein is localized to 11q12, among a cluster of related family members. Alternative splicing may result in multiple transcript variants; however, not all variants have been fully described.
Specifications
| MS4A3 | |
| Polyclonal | |
| Unconjugated | |
| MS4A3 | |
| CD20 antigen homolog; CD20 antigen-like protein; CD20L; hematopoietic cell 4 transmembrane protein; hematopoietic-specific transmembrane protein 4; HTM4; IgE receptor beta chain; IgE receptor beta subunit; membrane spanning 4-domains A3; membrane-spanning 4-domains subfamily A member 3; membrane-spanning 4-domains, subfamily A, member 3 (hematopoietic cell-specific); MS4A3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 932 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q96HJ5 | |
| MS4A3 | |
| Synthetic peptide located within the following region: AGPEELNTSV YQPIDGSPDY QKAKLQVLGA IQILNAAMIL ALGVFLGSLQ. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction