Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A4A Antibody (5C12), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312043AF750
Description
MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
MS4A4A | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
51338 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunofluorescence | |
5C12 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 | |
This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP. | |
0.1 mL | |
Primary | |
Human | |
IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction