Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A4A Mouse anti-Human, Clone: 5C12, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | MS4A4A |
---|---|
Clone | 5C12 |
Dilution | Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, ImmunoprecipitationSpecifications
MS4A4A | |
Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
RUO | |
PBS | |
51338 | |
IgG1 λ | |
Protein A purified |
5C12 | |
Monoclonal | |
Mouse | |
Human | |
4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 | |
This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title