Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107000 Supplier Invitrogen™ Supplier No. PA5107000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Human: 100%; Rat: 86%.

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. Alternative splicing of this gene results in several transcript variants; however, not all transcripts have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A4A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene MS4A4A
Gene Accession No. Q96JQ5
Gene Alias 4SPAN1; CD20 antigen-like 1; CD20L1; CD20-L1; Fc epsilon receptor beta subunit homolog; four-span transmembrane protein 1; HDCME31P; membrane spanning 4-domains A4A; membrane-spanning 4-domains subfamily A member 4A; membrane-spanning 4-domains, subfamily A, member 4; membrane-spanning 4-domains, subfamily A, member 4A; MS4A4; MS4A4A; MS4A7
Gene Symbols MS4A4A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human MS4A4A (aa 1-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.