Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A4A Recombinant Mouse Monoclonal Antibody (5C12)
GREENER_CHOICE

Catalog No. PIMA548222
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548222 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548222 Supplier Invitrogen™ Supplier No. MA548222
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: This antibody is specific for human extracellular MS4A4A.

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. Alternative splicing of this gene results in several transcript variants; however, not all transcripts have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A4A
Applications Flow Cytometry, Immunoprecipitation
Classification Recombinant Monoclonal
Clone 5C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene MS4A4A
Gene Accession No. Q96JQ5
Gene Alias 4SPAN1; CD20 antigen-like 1; CD20L1; CD20-L1; Fc epsilon receptor beta subunit homolog; four-span transmembrane protein 1; HDCME31P; membrane spanning 4-domains A4A; membrane-spanning 4-domains subfamily A member 4A; membrane-spanning 4-domains, subfamily A, member 4; membrane-spanning 4-domains, subfamily A, member 4A; MS4A4; MS4A4A; MS4A7
Gene Symbols MS4A4A
Host Species Mouse
Immunogen This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.