Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A4B Antibody (444008), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4504MFV500
Description
MS4A4B Monoclonal antibody specifically detects MS4A4B in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| MS4A4B | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 444008 | |
| Flow Cytometry, CyTOF-ready | |
| AI463180, Chandra, Ly116 | |
| HEK293 human embryonic kidney cell line transfected with mouse MS4A4B, Met1-Val226, Accession # NP_068364 | |
| RUO | |
| 60361 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction