Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A6A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282106
Description
MS4A6A Polyclonal specifically detects MS4A6A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MS4A6A | |
Polyclonal | |
Unconjugated | |
PBS | |
MS4A6A | |
MS4A6A antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MS4A6A. The immunogen is located within the first 50 amino acids of MS4A6A. Amino Acid Squence: SQAEKPEPTNQGQDSLKKH | |
0.1 mg | |
Primary | |
At least three isoforms of MS4A6A are known to exist; this antibody will detect the two longer isoforms. MS4A6A antibody is predicted to not cross-react with MS4A family members. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml | |
4SPAN3, CD20 antigen-like 3, CD20L3MS4A6A-polymorph, CD20-like precusor, Four-span transmembrane protein 3, four-span transmembrane protein 3.1,4SPAN3.2, four-span transmembrane protein 3.2, HAIRB-iso, membrane-spanning 4-domains subfamily A member 6A, membrane-spanning 4-domains, subfamily A, member 6A, MGC131944, MGC22650, MS4A6MST090, MSTP090 | |
Rabbit | |
Affinity Purified | |
RUO | |
64231 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction