Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A6A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548951
Description
MS4A6A Polyclonal Antibody for Western Blot
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
MS4A6A | |
Polyclonal | |
Unconjugated | |
MS4A6A | |
4SPAN3; 4SPAN3.2; CD20 antigen-like 3; CD20L3; CD20-like precusor; CDA01; four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; membrane spanning 4-domains A6A; membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; Ms4a11; MS4A6; MS4A6A; MS4A6A-polymorph; MST090; MSTP090 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
64231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9H2W1 | |
MS4A6A | |
KLH conjugated synthetic peptide between 11-38 amino acids from the N-terminal region of human MS4A6A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction