Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A6A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3083 Supplier Bioss Supplier No. BS13692R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in signal transduction as a component of a multimeric receptor complex.

Specifications

Antigen MS4A6A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MS4A6A
Gene Accession No. Q9H2W1
Gene Alias 4SPAN3; 4SPAN3.2; CD20 antigen-like 3; CD20L3; CD20-like precusor; CDA01; Four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; membrane spanning 4-domains A6A; Membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; Ms4a11; MS4A6; MS4A6A; MS4A6A-polymorph; MST090; MSTP090
Gene Symbols MS4A6A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MS4A6A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64231
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.