Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MS4A6E Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546038
Description
Peptide sequence: FILLSVNPAA LNPASLQCKL DEKDIPTRLL LSYDYHSPYT MDCHRAKASL Sequence homology: Human: 100%.
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members.
Specifications
MS4A6E | |
Polyclonal | |
Unconjugated | |
MS4A6E | |
membrane spanning 4-domains A6E; membrane-spanning 4-domains subfamily A member 6E; membrane-spanning 4-domains, subfamily A, member 6E; MS4A6E | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245802 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96DS6 | |
MS4A6E | |
Synthetic peptide directed towards the middle region of human MS4A6E. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction