Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A7 Antibody [CoraFluor™ 1], Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP347015CL1
Description
MS4A7 Polyclonal antibody specifically detects MS4A7 in Human samples. It is validated for ELISA,Western BlotSpecifications
| MS4A7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| CD20/FC-epsilon-RI-beta family member 4, CD20L4MGC22368, CFFM4CD20 antigen-like 4, Four-span transmembrane protein 2, high affinity immunoglobulin epsilon receptor beta subunit, membrane-spanning 4-domains subfamily A member 7, membrane-spanning 4-domains, subfamily A, member 7,4SPAN2, MS4A8 | |
| membrane-spanning 4-domains, subfamily A, member 7. (Uniprot# Q9GZW8) | |
| 0.1 mL | |
| Cell Biology | |
| 58475 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction