Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545837 Supplier Invitrogen™ Supplier No. PA545837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TGSLSIISGK QSTKPFDLSS LTSNAVSSVT AGAGLFLLAD SMVALRTASQ Sequence homology: Dog: 79%; Human: 100%.

This gene encodes a member of the membrane-spanning 4A gene family, members of which are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns in hematopoietic cells and nonlymphoid tissues. This family member is associated with mature cellular function in the monocytic lineage, and it may be a component of a receptor complex involved in signal transduction. This gene is localized to 11q12, in a cluster of other family members. At least four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MS4A7
Gene Accession No. Q9GZW8
Gene Alias 4SPAN2; CD20 antigen-like 4; CD20/Fc-epsilon-RI-beta family member 4; CD20L4; CFFM4; Four-span transmembrane protein 2; high affinity immunoglobulin epsilon receptor beta subunit; membrane spanning 4-domains A7; membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains subfamily A member 7; LOW QUALITY PROTEIN: membrane-spanning 4-domains subfamily A member 7; membrane-spanning 4-domains, subfamily A, member 7; MGC22368; MS4A7; MS4A8
Gene Symbols MS4A7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MS4A7.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58475
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.