Learn More
Invitrogen™ MS4A8B Monoclonal Antibody (1B4)

Description
Peptide Sequence: MNSMTSAVPV ANSVLVVAPH NGYPVTPGIM SHVPLYPNSQ PQVHLVPGNP PSLVSNVNGQ PVQKALKEGK TLGAIQIIIG LARIGLGSIM ATVLVGEYLS ISFYGGFPFW GGLWFIISGS LSVAAENQPY SYCLLSGSLG LNIVSAICSA VGVILFITDL SIPHPYAYPD YYPYAWGVNP GMAISGVLLV FCLLEFGIAC ASSHFGCQLV CCQSSNVSVI YPNIYAANPV ITPEPVTSPP SYSSEIQANK.
Specifications
Specifications
| Antigen | MS4A8B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1B4 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MS4A8 |
| Gene Accession No. | Q9BY19 |
| Gene Alias | 4SPAN4; CD20L5; Four-span transmembrane protein 4; membrane spanning 4-domains A8; membrane-spanning 4-domains subfamily A member 8; Membrane-spanning 4-domains subfamily A member 8B; membrane-spanning 4-domains, subfamily A, member 8; membrane-spanning 4-domains, subfamily A, member 8A; membrane-spanning 4-domains, subfamily A, member 8B; MS4A4; MS4A8; Ms4a8a; MS4A8B |
| Gene Symbols | MS4A8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.