Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552419 Supplier Invitrogen™ Supplier No. PA552419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNSMTSAVPV ANSVLVVAPH NGYPVTPGIM SHVPLYPNSQ PQVHLVPGNP PSLVSNVNGQ PVQKALKEGK Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 61%.

May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A8B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MS4A8
Gene Accession No. Q9BY19
Gene Alias 4SPAN4; CD20L5; Four-span transmembrane protein 4; membrane spanning 4-domains A8; membrane-spanning 4-domains subfamily A member 8; Membrane-spanning 4-domains subfamily A member 8B; membrane-spanning 4-domains, subfamily A, member 8; membrane-spanning 4-domains, subfamily A, member 8A; membrane-spanning 4-domains, subfamily A, member 8B; MS4A4; MS4A8; Ms4a8a; MS4A8B
Gene Symbols MS4A8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MS4A8B. Recombinant protein control fragment (Product #RP-88703).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.