Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587815 Supplier Invitrogen™ Supplier No. PA587815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: SISFYGGFPF WGGLWFIISG SLSVAAENQP YSYCLLSGSL GLNIVSAICS AVGVILFITD LSIPHPYAYP DYYPYAWGVN PGMAISGVLL VFCLLEFGIA C.

May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A8B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MS4A8
Gene Accession No. Q9BY19
Gene Alias 4SPAN4; CD20L5; Four-span transmembrane protein 4; membrane spanning 4-domains A8; membrane-spanning 4-domains subfamily A member 8; Membrane-spanning 4-domains subfamily A member 8B; membrane-spanning 4-domains, subfamily A, member 8; membrane-spanning 4-domains, subfamily A, member 8A; membrane-spanning 4-domains, subfamily A, member 8B; MS4A4; MS4A8; Ms4a8a; MS4A8B
Gene Symbols MS4A8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human MS4A8B (NP_1136451).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83661
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.