Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ CBRN Canister for Millennium and Ultra Elite Facepieces

Catalog No. 191300457 Shop All MSA Products
Change view
Click to view available options
For use with MSA Millennium 40mm threaded Gas Mask
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-130-0457 For use with MSA Millennium 40mm threaded Gas Mask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-0457 Supplier MSA™ Supplier No. 10046570
Add to Cart
Add to Cart

Canisters are compatible with MSA Millennium gas masks and MSA Ultra Elite CBRN gas masks.

  • NIOSH-approved when used with Millennium and Ultra Elite series gas masks
  • Gas mask canisters filter airborne contaminants for CBRN (chemical, biological, radiological, and nuclear) environments
  • Designed to protect against one or more specific hazards and are used with compatible headgear

Specifications

Cartridge Type CBRN
Certifications (NIOSH) NIOSH TC-14G-0254
Connection Type Threaded
Color Black
Product Type CBRN Canister
For Use With (Equipment) Millennium and Ultra Elite Gas Masks
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.