Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 1000 Riot Control Canisters, Pack of 6

Catalog No. 19042129 Shop All MSA Products
Change view
Click to view available options
For use with Advantage 1000 Gas Mask
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-042-129 For use with Advantage 1000 Gas Mask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-042-129 Supplier MSA™ Supplier No. 817590
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bayonet-type CBA/RCA canisters for use with Advantage 1000 and 3200 gas mask respirators.

  • NIOSH-approved when used with Advantage 1000 series gas masks
  • Gas mask canisters filter airborne contaminants for CBRN (chemical, biological, radiological, and nuclear) environments
  • Designed to protect against one or more specific hazards and are used with compatible headgear

Specifications

Cartridge Type Riot Control
Certifications (NIOSH) NIOSH TC-14G-0235
Connection Type Bayonet
Color Black
Product Type CBRN Canister
For Use With (Equipment) Advantage™ 1000 Gas Mask
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.