Learn More
MSA™ Advantage™ 1000 CBA-RCA / Millennium™ Gas Mask Accessory, Spectable Kit
Catalog No. 19041915 Shop All MSA Products
Change view
Click to view available options
Model:
816137
Catalog No. Model
19-041-915 816137
1 options
Catalog No. 19-041-915 Supplier MSA™ Supplier No. 816137
Add to Cart
Add to Cart
Gas mask accessory for use with Advantage or Millenium facepieces.
  • Spectacle Kit adapts corrective lenses for use with facepiece and includes wire support, rubber guide, and one pair of 44mm lens-size, metal-frame spectacles (minus lenses)
Specifications
Product Type Kit
For Use With (Equipment) Advantage 1000 CBA-RCA, Millennium Gas Mask
Includes Wire support, rubber guide and one pair of 44 mm frames (without lenses)
Model 816137