Learn More
Description
Requires
Advantage cartridges and filters (sold separately). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.
Low-maintenance, weighs 40% less than conventional full-face respirators. Constructed of soft Hycar™™ rubber that provides the feel and comfort of silicone but with better permeation resistance.
- Fully elastic, six-point head harness adjusts easily
- Interchangeable Advantage Series cartridges enhance versatility and are compatible with Advantage 200, Advantage 420, Advantage 4000 and Advantage 3200 respirators
- Unique inturned lip seal on facepiece provides an exceptional seal against a wide range of facial contours
- Flexible urethane lens is integrally bonded to the facepiece, eliminating the need for rigid lens retainers
- Includes a speaking diaphragm for clear short-range communications and a nosecup to reduce lens fogging in low temperatures or high humidity
Order Info
Optional lens cover, lens shields, and ESP™ communication system available separately.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| Certifications/Compliance | AS NZS 1716:2003 |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Color | Black |
| Material | Hycar Rubber |
| Product Type | Full Facepiece Respirator |
| Size | Small |
| Facepiece Material | Hycar™ |
| For Use With (Application) | Fire Service, general industry, oil and gas, utilities |
| For Use With (Equipment) | Advantage™ Series cartridges |
| Harness Material | Elastic |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.