Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 200 LS Half-Mask Respirators

Catalog No. 19039023A Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
Neck Strap:
Single
Two-piece
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Neck Strap
19-039-023A Small Two-piece
18-999-3899 Small Single
18-999-3897 Medium Single
18-999-3898 Large Single
19-039-023B Medium Two-piece
19-039-023C Large Two-piece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Cartridges and filters sold separately
Catalog No. 19-039-023A Supplier MSA™ Supplier No. 815696
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cartridges and filters sold separately

Requires

Advantage cartridges and filters (see separate listings). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.

Half-mask respirators offer maximum protection and high wearing comfort with its patented MultiFlex system. Ideal for applications where workers are exposed to various hazards such as high concentrations of fumes, mists and gases. Available in single or two-piece neckstrap.

  • Comfortable, efficient and economic half mask
  • Light, soft pliable thermoplastic facepiece
  • Multiflex&;trade; System equalizes pressure on the face-seal area while delivering stability
  • Available with single or two-piece neck strap
  • Ideal for applications where workers are exposed to high concentrations of fumes, mists and gases
  • Customized and pressure-free fit, instantly conforms to the wearer’s unique facial characteristics
  • Available in three sizes and has a full range of cartridges and filters to protect against harmful particles and gases

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Certifications (EN) EN 140
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
For Use With (Equipment) Advantage™ Flexi-Filters™ and Advantage™ Cartridges
Model 815696
Neck Strap Two-piece
Suspension Type 4-point
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.