Learn More
Description
Requires
Advantage cartridges and filters (see separate listings). Also compatible with Comfo Series cartridges when used with adapter 18-999-1504.
- Comfortable, efficient and economic half mask
- Light, soft pliable thermoplastic facepiece
- Multiflex&;trade; System equalizes pressure on the face-seal area while delivering stability
- Available with single or two-piece neck strap
- Ideal for applications where workers are exposed to high concentrations of fumes, mists and gases
- Customized and pressure-free fit, instantly conforms to the wearer’s unique facial characteristics
- Available in three sizes and has a full range of cartridges and filters to protect against harmful particles and gases
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| Certifications (EN) | EN 140 |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Color | Blue |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| Size | Medium |
| For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
| For Use With (Equipment) | Advantage™ Flexi-Filters™ and Advantage™ Cartridges |
| Model | 815692 |
| Neck Strap | Two-piece |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.