Learn More
Description
Requires
NIOSH-approved Advantage series cartridges and filters.
- Dual-port has lower breathing resistance than traditional full-facepiece respirators
- Soft, pliable silicone facemask and rubber head harness don quickly
- Pre-adjustable straps ensure a consistent fit and tighter seal
- Optically correct, wraparound facepiece lens provides a wide, distortion-free field of vision
- Scratch-resistant lens coating
- Fog-reducing standard nosecup
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| ANSI Standard | ANSI Z87.1 |
| Certifications/Compliance | AS NZS 1716:2003 |
| Certifications (EN 136) | Class 2 |
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Connection Type | Bayonet |
| Color | Blue |
| Material | Silicone |
| Product Type | Full Facepiece Respirator |
| Size | Large |
| Facepiece Material | Silicone |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.