Learn More
MSA™ Advantage™ 420 Half-Mask Respirator
Catalog No. 17200084 Shop All MSA Products
Change view
Click to view available options
Model:
Advantage 420 Respirator Assembly
Advantage 420 Respirator Threaded Cartridge Assembly
Includes:
Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve
Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters
Size:
Small
Medium
Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes Size
17-200-084 Advantage 420 Respirator Threaded Cartridge Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters Large
19-825-001 Advantage 420 Respirator Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve Medium
19-825-000 Advantage 420 Respirator Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve Small
19-825-002 Advantage 420 Respirator Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve Large
17-200-085 Advantage 420 Respirator Threaded Cartridge Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters Medium
17-200-086 Advantage 420 Respirator Threaded Cartridge Assembly Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-200-084 Supplier MSA™ Supplier No. 10119574
Add to Cart
Add to Cart
One-piece UniBond over-molded silicone facepiece helps eliminate possible leak paths
  • Drop-down harness position allows respirator to be removed while retaining head protection
  • One-piece ComforTop™ cradle headband eliminates pressure points
  • AnthroCurve™ multi-ethnic face seal improves fit-testing among diverse workforce
  • Lock-down harness position allows a personal fit without further user adjustments
  • No loose or dangling straps to impede operation or vision
  • Latex-free; high-quality silicone provides greater durability and product life
  • Textured inner facepiece seal minimizes slippage, improves fit
  • Bayonet and threaded cartridge options allow use of all MSA cartridges
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
Meets OSHA 29 CFR Part 1920.134
Specifications
Certifications/Compliance NIOSH 42CFR84
Material Silicone
Product Type 420 Half Mask Respirator Assembly
Size Large
For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
For Use With (Equipment) Respiratory Systems
Includes Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve, 2 threaded cartridge adapters
Latex Free Yes
Model Advantage 420 Respirator Threaded Cartridge Assembly