Description
- Drop-down harness position allows respirator to be removed while retaining head protection
- One-piece ComforTop™ cradle headband eliminates pressure points
- AnthroCurve™ multi-ethnic face seal improves fit-testing among diverse workforce
- Lock-down harness position allows a personal fit without further user adjustments
- No loose or dangling straps to impede operation or vision
- Latex-free; high-quality silicone provides greater durability and product life
- Textured inner facepiece seal minimizes slippage, improves fit
- Bayonet and threaded cartridge options allow use of all MSA cartridges
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
Meets OSHA 29 CFR Part 1920.134
Specifications
Specifications
| Certifications/Compliance | NIOSH, AS/NZS 1716:2003, 42 CFR 84 |
| Material | Silicone |
| Product Type | Half Mask Respirator |
| Size | Small |
| For Use With (Application) | Asbestos abatement, painting, sanding, grinding, welding |
| For Use With (Equipment) | Respiratory Systems |
| Includes | Facepiece, head cradle, elastic straps, buckle set, yoke with lever, 2 inhalation valves, 1 exhalation valve |
| Latex Free | Yes |
| Model | Advantage 420 Respirator Assembly |