Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ Cartridges

Catalog No. 189993919 Shop All MSA Products
Change view
Click to view available options
Cartridge Type:
Acid Gas Cartridge
Ammonia, Methylamine Cartridge
Low Profile Particulate Cartridge
Mercury Vapor Cartridge
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge
Organic Vapor, Chlorine, Sulfur Dioxide, Chlorine Dioxide, Hydrogen chloride, Hydrogen sulfide, Ammonia, Methylamine, Formaldehyde, Hydrogen fluoride, Particulates Cartridge
Filter Type:
GMA-P100 Filter
GMI-P100 Filter
N95, R951
P100
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Filter Type
18-999-3919 Organic Vapor, Acid Gas Cartridge N95, R951
18-999-3765 Mercury Vapor Cartridge N95, R951
18-999-3808 Acid Gas Cartridge P100
18-999-3810 Organic Vapor Cartridge GMA-P100 Filter
18-999-3920 Acid Gas Cartridge N95, R951
18-999-3923 Organic Vapor Cartridge N95, R951
18-999-3978 Low Profile Particulate Cartridge P100
19-003-230 Organic Vapor, Chlorine, Sulfur Dioxide, Chlorine Dioxide, Hydrogen chloride, Hydrogen sulfide, Ammonia, Methylamine, Formaldehyde, Hydrogen fluoride, Particulates Cartridge P100
19-033-002B Organic Vapor, Acid Gas, Ammonia, Methylamine, Formaldehyde, Hydrogen Fluoride Cartridge N95, R951
19-033-002C Organic Vapor Cartridge GMI-P100 Filter
18-999-3791 Ammonia, Methylamine Cartridge P100
18-999-3807 Organic Vapor, Acid Gas Cartridge P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 18-999-3919 Supplier MSA™ Supplier No. 815357

Lightweight for comfort, but designed for maximum protection

  • Particulate, chemical, and combination cartridges
  • Bayonet-style mounting allows easy installation—cartridges lock into place with a single twist
  • Interchangeable between Advantage 200LS, Advantage 420, Advantage 1000, Advantage 4000 and Advantage 3200 respirators

Order Info

Packaged two cartridges per pack.

Compliance

NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirements; EN 14387. Meets OSHA 29 CFR Part 1920.134.

Specifications

Cartridge Type Organic Vapor, Acid Gas Cartridge
Certifications/Compliance OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU
Connection Type Bayonet
Color Yellow
Filter Type N95, R951
Product Type Advantage™ Respirator Cartridge
For Use With (Application) Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue
For Use With (Equipment) Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series)
Quantity 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.