Learn More
Description
- Particulate, chemical, and combination cartridges
- Bayonet-style mounting allows easy installation—cartridges lock into place with a single twist
- Interchangeable between Advantage 200LS, Advantage 420, Advantage 1000, Advantage 4000 and Advantage 3200 respirators
Order Info
Packaged two cartridges per pack.
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirements; EN 14387. Meets OSHA 29 CFR Part 1920.134.
Specifications
Specifications
| Cartridge Type | Organic Vapor Cartridge |
| Certifications/Compliance | OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU |
| Connection Type | Bayonet |
| Color | Black |
| Filter Type | N95, R951 |
| Product Type | Advantage™ Respirator Cartridge |
| For Use With (Application) | Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue |
| For Use With (Equipment) | Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series) |
| Quantity | 2/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.