Learn More
MSA™ Air-Supply Hoses for Airline Respirators
Catalog No. 189993296 Shop All MSA Products
Catalog No. 18-999-3296 Supplier MSA™ Supplier No. 455022
Add to Cart
Add to Cart
For use with PremAire supplied air respirators.
Temperature Ranges
  • Neoprene: +32 to +100°C (-25 to +212°F)
  • PVC: 0 to 49°C (32 to 120°F)
  • Coiled Nylon: 18 to 71°C (0 to 160°F)
An MSA air-supply hose must be used to maintain NIOSH/MSHA approvals of the PremAire respirator
Specifications
Model 455022