Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ Pro Single Gas Detector

Catalog No. 19815621 Shop All MSA Products
Change view
Click to view available options
Quantity:
Bulk Pack
Single Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-815-621 Bulk Pack
19-810-243 Single Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-815-621 Supplier MSA™ Supplier No. 10096703

missing translation for 'validMainframeMsg'

Provides worry-free performance, standing up to the roughest handling in tough industrial environments

  • Built-in gas delivery adapter; no accessories are needed for gas response tests
  • Distinctive triple alarm system leaves no doubt for users in alarm situations
  • Extended frequency range covers a wide sound spectrum
  • Visual alarm features dual LED displays located on the unit's top, visible from all angles
  • Internal vibrating alarm is standard, emitting a pulse that directly calls additional attention when in alarm mode

Detects Hydrogen Sulfide

Certifications

Class I, Div. 1, Groups A, B, C and D; Class II, Div. 1, Group G, AEx ia IIC T4

Warranty and Services

Two Years from date of purchase

Specifications

Components Measured Hydrogen Sulfide
Type Single Gas Detector
Quantity Bulk Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.