Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ Pro Single Gas Detector: O-Rings

Catalog No. 19815628 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Altair Pro Single Gas Detector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-815-628 Altair Pro Single Gas Detector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-815-628 Supplier MSA™ Supplier No. 635511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for Altair Pro Single Gas Detector.

  • For O2-remote version cable
  • Two required

Specifications

Color Black
For Use With (Equipment) Altair Pro Single Gas Detector
Type O-ring
Material Buna-N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.