Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ Pro Single-Gas Detectors (Alternate Setpoint Models)

Catalog No. 19165571 Shop All MSA Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide
Hydrogen Sulfide
Oxygen
Measurement Range:
0 to 1500
0 to 200
0 to 25%
Resolution:
0.1%
1
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Resolution
19-165-571 Hydrogen Sulfide 0 to 200 1
19-165-563 Oxygen 0 to 25% 0.1%
19-165-564 Carbon Monoxide 0 to 1500 1
19-165-569 Hydrogen Sulfide 0 to 200 1
19-165-570 Hydrogen Sulfide 0 to 200 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-165-571 Supplier MSA™ Supplier No. 10076726

missing translation for 'validMainframeMsg'

Provides worry-free performance and stands up to the roughest handling in even the toughest industrial environments

These detectors accurately measures and display gas concentration or percent oxygen on a large, clear, backlit LCD. They offer excellent impact resistance and RFI performance.

  • Tough rubberized housing
  • One-button operation
  • Superior dust and water protection (rated IP67 except O2-R)
  • Event-logging and data-logging
  • Adjustable alarm set points are offered for LOW, HIGH, TWA, and STEL
  • Alarms are indicated by flashing LEDs, audible alarm, and internal vibrating alarm
  • Replaceable battery and sensor

  • Chemical
  • Construction
  • Electric Utility
  • Firefighting
  • Gas Detection
  • General Industry
  • Hazardous Materials
  • Indoor Air Quality
  • Manufacturing
  • Nuclear
  • Oil and Gas
  • Water/Wastewater

Certifications

Class I, Div. 1, Groups A, B, C and D; Class II,

Div. 1, Group G, AEx ia IIC T4

Warranty and Services

2 years from date of purchase

Specifications

Alarm Types Audible, visual, vibrating
Components Measured Hydrogen Sulfide
Dimensions (L x W x H) 3.4 x 2 x 1 in. (8.63 x 5.08 x 2.54cm)
Height (English) 1 in.
Height (Metric) 2.54 cm
Humidity Range 10 to 95%
IP Rating IP67
Length (English) 3.4 in.
Length (Metric) 8.63 cm
Measurement Range 0 to 200
Resolution 1
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 2 Years
Weight (English) 4 oz.
Weight (Metric) 113g
Width (English) 2 in.
Width (Metric) 5.08 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.