Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ Single-Gas Detector

Catalog No. p-3574163 Shop All MSA Products
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
Oxygen
Measurement Range:
0 to 100
0 to 25%
Resolution:
0.1%
1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Resolution
19-150-0381 Hydrogen Sulfide 0 to 100 1
17-200-058 Oxygen 0 to 25% 0.1%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-0381 Supplier MSA™ Supplier No. 10092521
Only null left
Add to Cart
Add to Cart

Includes

Rugged suspension clip

Factory-calibrated detector offers two-year, maintenance-free service life

Unit will not deactivate after two years of operation, but will continue to operate as long as battery capacity and sensor performance allow.

  • Easy-to-use instrument features one-button operation with no required sensor or battery changes
  • Extremely small and thin sensor profile allows unit to deliver high performance in a small package
  • Large numeric display allows user to view service life countdown, alarm conditions, setpoints, peak readings, and hours in alarm since activation
  • Triple alarm system consists of piercing 95dB audible alarm, dual super-bright LED alarm lights, and a pulse-emitting vibrating alarm
  • Bump check is confirmed with checkmark on instrument LCD; displays for 24 hours
  • Event recorder stores information about 25 latest events automatically
  • Extra-thick rubberized housing resists impact and water/dust ingress
  • Built-in IR communications capability allows user to download and analyze readings as well as adjust alarm setpoints
  • Functions in a temperature range of -4° to +122°F (-20° to +50°C); relative humidity, 10 to 95%

Order Info

Dimensions (L x W x H): 1 x 2 x 3.4 in. (2.5 x 5.1 x 8.6cm). Weight: 4 oz. (113g) including clip.

Certifications

Rugged housing is IP67 rated for ingress protection. Classified as Intrinsically Safe for use in Class I, Division 1, Groups A, B, C, and D hazardous locations in the U.S. and Canada; ATEX, II 2 G EEx ia IIC T4.

Warranty and Services

Two years or 1080 alarm minutes (18 hours) under normal conditions.

Specifications

Alarm Types Audible, visual, vibrating
Components Measured Hydrogen Sulfide
Dimensions (L x W x H) 3.4 x 2 x 1 in. (8.63 x 5.08 x 2.54cm)
Height (English) 1 in.
Height (Metric) 2.54 cm
Humidity Range 10 to 95%
IP Rating IP67
Length (English) 3.4 in.
Length (Metric) 8.63 cm
Measurement Range 0 to 100
Resolution 1
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 2 Years
Weight (English) 4 oz.
Weight (Metric) 113g
Width (English) 2 in.
Width (Metric) 5.08 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.