Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Battery Pack for ALTAIR™ Multigas Detector

Catalog No. 17200046 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Altair 5X Automated Gas Detector
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-200-046 Altair 5X Automated Gas Detector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-046 Supplier MSA™ Supplier No. 10114837
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Battery Pack, alkaline for ALTAIR multigas detectors

For use with ALTAIR multigas detectors

Specifications

Color Black
For Use With (Equipment) Altair 5X Automated Gas Detector
Type Alkaline Battery Pack
Includes Belt clip
Voltage 6.7VDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.