Learn More
MSA™ Bosun™ Chair
Catalog No. 191303876 Shop All MSA Products
Change view
Click to view available options
Certifications/Compliance:
OSHA, ANSI, CSA
Catalog No. Certifications/Compliance
19-130-3876 OSHA, ANSI, CSA
1 options
Catalog No. 19-130-3876 Supplier MSA™ Supplier No. SSH3505017
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suspension seat for lifting and lowering personnel
  • For hands-free work operations in seated position
  • With padded plastic seat
  • Use with full-body harnesses to sit suspended for long durations
  • Attached to chest D-Ring
Specifications
Material Aluminum
Certifications/Compliance OSHA, ANSI, CSA
Product Type Bosn's chair
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.