Learn More
MSA™ Cairns™ Invader 664 Fire Helmets: ESS Goggle System
Catalog No. 191302394 Shop All MSA Products
Change view
Click to view available options
Liner Material:
Flannel Cushion
Leather Cushion
Color:
Black
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Liner Material Color
19-130-2394 Flannel Cushion Yellow
19-130-2395 Flannel Cushion White
19-130-2397 Flannel Cushion Red
19-130-2396 Flannel Cushion Black
19-130-2408 Leather Cushion Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-130-2394 Supplier MSA™ Supplier No. 0197114511F
Solid protection and comfort
Duraglas™ thermoplastic composite material resists cracking and chipping.
  • Flame-resistant earflap and Nomex™ chin strap increase protection
  • SCBA interface front headband adjustment matches the fit of any SCBA mask
  • Three position rear ratchet height adjustment ensures secure fit for all head sizes between 16.2 and 21.3cm (6.38 and 8.38 in.)
  • High temperature foam impact cap and liner offer higher protection against impact
Standard Configuration
  • Flannel cushion liner
  • Nomex earflap
Deluxe Configuration
  • Leather cushion liner
  • Flannel crown pad
  • PBI/Kevlar™ earflaps
  • Defender visor
Certifications
NFPA 1971-2007 certified
Specifications
Color Yellow
Liner Material Flannel Cushion