Learn More
MSA™ Cairns™ Invader 664 Fire Helmets: ESS Goggle System
Catalog No. 191302408 Shop All MSA Products
Click to view available options
Liner Material:
Flannel Cushion
Leather Cushion
Color:
Black
Red
White
Yellow
Solid protection and comfort
Duraglas™ thermoplastic composite material resists cracking and chipping.
  • Flame-resistant earflap and Nomex™ chin strap increase protection
  • SCBA interface front headband adjustment matches the fit of any SCBA mask
  • Three position rear ratchet height adjustment ensures secure fit for all head sizes between 16.2 and 21.3cm (6.38 and 8.38 in.)
  • High temperature foam impact cap and liner offer higher protection against impact
Standard Configuration
  • Flannel cushion liner
  • Nomex earflap
Deluxe Configuration
  • Leather cushion liner
  • Flannel crown pad
  • PBI/Kevlar™ earflaps
  • Defender visor
Certifications
NFPA 1971-2007 certified
Specifications
Color Black
Liner Material Leather Cushion
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.