Learn More
MSA™ Cairns™ Invader 664 Fire Helmets: Tuffshield
Catalog No. 191302386 Shop All MSA Products
Change view
Click to view available options
Liner Material:
Flannel Cushion
Leather Cushion
Faceshield Material:
4 in. Tuffshield
Cairns Goggle System
Color:
Black
Red
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Liner Material Faceshield Material Color
19-130-2386 Flannel Cushion 4 in. Tuffshield Yellow
19-130-2388 Flannel Cushion 4 in. Tuffshield Black
19-130-2387 Flannel Cushion 4 in. Tuffshield White
19-130-2389 Flannel Cushion 4 in. Tuffshield Red
19-130-2398 Leather Cushion 4 in. Tuffshield Yellow
19-130-2399 Leather Cushion 4 in. Tuffshield White
19-130-2400 Leather Cushion 4 in. Tuffshield Black
19-130-2390 Flannel Cushion Cairns Goggle System Yellow
19-130-2391 Flannel Cushion Cairns Goggle System White
19-130-2392 Flannel Cushion Cairns Goggle System Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-130-2386 Supplier MSA™ Supplier No. 664FSY
Add to Cart
Add to Cart
Solid protection and comfort
Duraglas™ thermoplastic composite material resists cracking and chipping.
  • Flame-resistant earflap and Nomex™ chin strap increase protection
  • SCBA interface front headband adjustment matches the fit of any SCBA mask
  • Three position rear ratchet height adjustment ensures secure fit for all head sizes between 16.2 and 21.3cm (6.38 and 8.38 in.)
  • High temperature foam impact cap and liner offer higher protection against impact
Standard Configuration
  • Flannel cushion liner
  • Nomex earflap
Deluxe Configuration
  • Leather cushion liner
  • Flannel crown pad
  • PBI/Kevlar™ earflaps
  • Defender visor
Certifications
NFPA 1971-2007 certified
Specifications
Color Yellow
Faceshield Material 4 in. Tuffshield
Liner Material Flannel Cushion