Description
Duraglas™ thermoplastic composite material resists cracking and chipping.
- Flame-resistant earflap and Nomex™ chin strap increase protection
- SCBA interface front headband adjustment matches the fit of any SCBA mask
- Three position rear ratchet height adjustment ensures secure fit for all head sizes between 16.2 and 21.3cm (6.38 and 8.38 in.)
- High temperature foam impact cap and liner offer higher protection against impact
Standard Configuration
- Flannel cushion liner
- Nomex earflap
- Leather cushion liner
- Flannel crown pad
- PBI/Kevlar™ earflaps
- Defender visor
Certifications
NFPA 1971-2007 certified
Specifications
Specifications
| Color | White |
| Faceshield Material | 4 in. Tuffshield |
| Liner Material | Flannel Cushion |