Learn More
MSA™ Cap Adapters
Catalog No. 189993844 Shop All MSA Products
Change view
Click to view available options
Product Type:
Positive Positioning Adapter Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-999-3844 Positive Positioning Adapter Kit
18-999-3746 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3844 Supplier MSA™ Supplier No. 806211
Specifications
For Use With (Equipment) Sparkgards and Welding Shields to Slotted Cap
For Use With (Application) Welding
Product Type Positive Positioning Adapter Kit