Description
Includes
Mask comes with facepiece and canister.
- Face and respiratory protection against particulates, vapors, and gases
- Ultravue™ facepiece constructed of black Hycar™™ rubber with wraparound poly lens
- Lightweight, replaceable canister
- Small, oval-shaped aluminum canister for easy downward head movement
- Inhalation check valve prevents exhaled air from entering breathing tube
- Mechanical speaking diaphragm for short-range communication
- Five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position
- Maximum Use Conditions:
- Entry, 100 times the Permissible Exposure Level (PEL) without exceeding the Immediately Dangerous to Life or Health (IDLH) concentration
- Escape, contaminant concentrations up to 5000 ppm (0.5% by volume) providing presence of adequate oxygen (19.5% by volume)
Certifications
Canisters are NIOSH approved.
Specifications
Specifications
| Cartridge/Filter | Chlorine |
| Color | Black |
| Product Type | Gas Mask Respirator |
| Model | 817065 |