Learn More
MSA™ Chin-Type Gas Masks
Catalog No. 19033000H Shop All MSA Products
Change view
Click to view available options
Cartridge/Filter:
Ammonia
Chlorine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge/Filter
19-033-000H Chlorine
19-033-000F Ammonia
19-033-000G Chlorine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-033-000H Supplier MSA™ Supplier No. 817066
Includes
Mask comes with facepiece and canister.
Chin-type full face respirators provide protection against particulates, vapors, and gases by way of Ultravue™ facepiece and lightweight, replaceable canister.
  • Face and respiratory protection against particulates, vapors, and gases
  • Ultravue™ facepiece constructed of black Hycar™™ rubber with wraparound poly lens
  • Lightweight, replaceable canister
  • Small, oval-shaped aluminum canister for easy downward head movement
  • Inhalation check valve prevents exhaled air from entering breathing tube
  • Mechanical speaking diaphragm for short-range communication
  • Five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position
  • Maximum Use Conditions:
    • Entry, 100 times the Permissible Exposure Level (PEL) without exceeding the Immediately Dangerous to Life or Health (IDLH) concentration
    • Escape, contaminant concentrations up to 5000 ppm (0.5% by volume) providing presence of adequate oxygen (19.5% by volume)
Certifications
Canisters are NIOSH approved.
Specifications
Cartridge/Filter Chlorine
Color Black
Product Type Gas Mask Respirator
Model 817066